Learn More
Waters Corp Needle Kit, FEP/Metal, 20μL, I-Class
Supplier:  Waters Corp 700005929
Encompass_Preferred
This needle kit provides an optional 20μL FEP/Metal needle assembly. It is designed for use in ACQUITY I-Class Fixed Loop (FL) Sample Managers and UPLC systems.
  • Optional 20μL FEP/Metal needle assembly.
  • Designed for use in ACQUITY I-Class Fixed Loop (FL) Sample Managers.
  • Recommended annual replacement.
Catalog No. 50-239-1177