Learn More
Waters Corp Waters Corporation Needle Kit, I-Class, PEEKsil, 10 μL
Catalog No. 502039561 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This optional 10μL PEEKsil needle assembly is designed for use in ACQUITY I-Class Fixed Loop (FL) Sample Managers.
  • Provided as a spare parts kit.
  • One unit per package.
  • Compatible with UPLC systems.
  • Recommended annual replacement.
Catalog No. 50-203-9561 Supplier Waters Corp Supplier No. 700005926
This optional 10μL PEEKsil needle assembly is designed for use in ACQUITY I-Class Fixed Loop (FL) Sample Managers.
  • Provided as a spare parts kit.
  • One unit per package.
  • Compatible with UPLC systems.
  • Recommended annual replacement.