Learn More
Waters Corp Waters Needle Kit, I-Class, Steel, 10 μL
Catalog No. 502039559 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Kit, I-Class Steel Needle, 10 μL is an optional stainless steel needle assembly for use in Waters I-Class Fixed Loop (FL) Sample Managers. It is particularly beneficial when the system is updated for Hexane/THF solvent compatibility.
  • Optional 10 μL stainless steel needle assembly.
  • Designed for use in I-Class Fixed Loop (FL) Sample Managers.
  • Especially useful for systems updated for Hexane/THF solvent compatibility.
Catalog No. 50-203-9559 Supplier Waters Corp Supplier No. 700005924
The Kit, I-Class Steel Needle, 10 μL is an optional stainless steel needle assembly for use in Waters I-Class Fixed Loop (FL) Sample Managers. It is particularly beneficial when the system is updated for Hexane/THF solvent compatibility.
  • Optional 10 μL stainless steel needle assembly.
  • Designed for use in I-Class Fixed Loop (FL) Sample Managers.
  • Especially useful for systems updated for Hexane/THF solvent compatibility.