The SpectraMax iD5 is a five-mode multi-mode microplate reader designed for routine and advanced laboratory assays, combining monochromator and filter optics with an intuitive touchscreen and SoftMax software to support absorbance, fluorescence, luminescence, TRF, and FP workflows.
- Five detection modes: absorbance, fluorescence intensity, luminescence, time-resolved fluorescence, fluorescence polarization.
- Plate formats: compatible with 6 to 384 well microplates.
- Optics: hybrid monochromator and filter-based reads with xenon flash lamp.
- Detectors: photomultiplier tube and photodiode.
- Temperature control: 5 °C above ambient to 66 °C.
- Shaking modes: linear, orbital, double orbital.
- Software: embedded SoftMax Touch interface and SoftMax Pro compatibility for data analysis.
- Dimensions and weight: 15.79 x 20.94 x 23.54 in (40.1 x 53.2 x 59.8 cm); 88.1 lb (40 kg).