Learn More
Grainger Blodgett Temperature Control Kit, Digital Timer, 150°F to 500°F, 1 ea
Catalog No. 502795203
Encompass_Preferred
This infinite control kit with digital timer is designed for use with select Blodgett convection ovens, facilitating precise temperature regulation and consistent cooking cycles. The kit includes all necessary components for installation and integrates directly with specified models to enhance operational control.
  • Provides precise temperature control within a range of 150°F to 500°F
  • Features a 60-minute digital timer with an integrated audible alarm
  • Includes an infinite switch, mounting bracket, and required installation hardware
  • Designed to fit a standard 1/2 in. mounting hole
  • Utilizes UL listed components
  • Compatible with DFG-100, DFG-200, Mark V, and Mark V-100 series convection ovens
Catalog No. 50-279-5203 Supplier Grainger Supplier No. 19XL06
This infinite control kit with digital timer is designed for use with select Blodgett convection ovens, facilitating precise temperature regulation and consistent cooking cycles. The kit includes all necessary components for installation and integrates directly with specified models to enhance operational control.
  • Provides precise temperature control within a range of 150°F to 500°F
  • Features a 60-minute digital timer with an integrated audible alarm
  • Includes an infinite switch, mounting bracket, and required installation hardware
  • Designed to fit a standard 1/2 in. mounting hole
  • Utilizes UL listed components
  • Compatible with DFG-100, DFG-200, Mark V, and Mark V-100 series convection ovens