Learn More
Med Vet International Miltex Meisterhand General Surgery Kit Without Cassette, Mh6800-Nc
Supplier:  Med Vet International MILMH6800NC
Encompass_Preferred
The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. This is a Miltex MeisterHand General Surgery Kit. Specifications: Without Cassette, MH6800-NCContents(1) Mayo Scissors, Straight, 6-3/4" (173 mm) (1) #3 Knife H and le(1) Snook Ovariectomy Hook, 8" (203 mm) (3) Halsted Mosquito Forceps, Curved, 5" (122 mm) (2) Kelly Forceps, Straight, 5-1/2" (140 mm) (1) Metzenbaum Scissors, Curved, 7" (178 mm) (1) Brown-Adson Forceps, 7 x 7 Teeth, 4-3/4" (121 mm) (4) Rochester-Carmalt Forceps, Curved, 6-1/4" (159 mm) (4) Backhaus Towel Clamps, 5-1/4" (133 mm) (1) Olsen-Hegar Needle Holder, Tungsten Carbide, Serrated 5-1/2" (140 mm) (1) Vet General Surgical Cassette, 15" x 8-1/4" x 1-1/4"
Catalog No. 50-283-2116