Learn More
TA Instruments Kit MDSC Calibration Sapphire, Two Sapphire Discs, MDSC Calibration of DSC, 0.160 Inch Disc for Standard Pans, 0.135 Inch Disc for Hermetic Pans
Catalog No. 5000580893 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Kit MDSC Calibration Sapphire contains two sapphire discs for MDSC calibration of the DSC instrument. Includes 0.160 inch OD x 0.020 inch height disc for standard sample pans and 0.135 inch OD x 0.025 inch height disc for hermetic sample pans.
Catalog No. 50-005-80893 Supplier TA Instruments Supplier No. 970370.901
Waters TA Instruments Kit MDSC Calibration Sapphire contains two sapphire discs for MDSC calibration of the DSC instrument. Includes 0.160 inch OD x 0.020 inch height disc for standard sample pans and 0.135 inch OD x 0.025 inch height disc for hermetic sample pans.