Learn More
Med Vet International 38-Pc. First Aid Medical Kit For Pets - Advanced Edition
Supplier:  Med Vet International SHS4
Encompass_Preferred
This advanced first aid medical kit for pets contains everything you need to treat minor to moderate wounds, including wound closure supplies such as a sterile skin stapler preloaded with 35 staples. This comprehensive kit comes with a variety of supplies to treat your pet in an emergency. From the dog park to extended hunting trips, this kit will keep you prepared for your pet’s safety. Contents: (1) 2in Adson Forceps(4) 4in x 4.1yds Conforming Stretch Gauze Roll(2) Medium Nitrile Gloves(1) 1in Blood Stop Hemostatic Gauze(8) 6in Cotton Tipped Applicators(4) 1"x1.5yds Plastic Tape(4) 2x3in Non-Adherent Dressing(4) 2in Elastic Bandages(4) Antiseptic Towelette(1) Triangle Bandage With Safety Pins(1) 7 3/4in Utility Bandage Scissors(1) Instant Cold Pack Compress(2) 1.5in Safety Pins(1) Sterile Skin Stapler All instruments are medical grade stainless steel. Skin Stapler is sterile. When it counts, this lightweight and compact first aid kit will be easily deployable to treat most wounds.
Catalog No. 50-282-9939