Learn More
Chemglass Life Sciences Kit, Minum-Ware, Basic
SDP
Catalog No. 50873337
Encompass_Preferred
Kit, Minum-Ware, Basic. This kit contains the microscale components to do the majority of experiments described in Organic Laboratory Techniques, Donald L. Pavia, Gary M. Lampman, George S. Kriz, Randy G. Engel, Sanders College Publishing-Chicago IL. and Microscale Organic Laboratory, Dana W. Mayo, Ronald M. Pike, Samuel S. Butcher, John S. Wiley & Sons, NY., NY. The MW-16-01 kit is packaged in a polypropylene box having movable dividers permitting additional components to be added at a later date. All joints are 14/10 with the outer members externally threaded. Large quantity discounts are available, please call for details. For a complete list of components, see the specifications tab below.
Catalog No. 50-873-337 Supplier Chemglass Life Sciences Supplier No. MW1601
Kit, Minum-Ware, Basic. This kit contains the microscale components to do the majority of experiments described in Organic Laboratory Techniques, Donald L. Pavia, Gary M. Lampman, George S. Kriz, Randy G. Engel, Sanders College Publishing-Chicago IL. and Microscale Organic Laboratory, Dana W. Mayo, Ronald M. Pike, Samuel S. Butcher, John S. Wiley & Sons, NY., NY. The MW-16-01 kit is packaged in a polypropylene box having movable dividers permitting additional components to be added at a later date. All joints are 14/10 with the outer members externally threaded. Large quantity discounts are available, please call for details. For a complete list of components, see the specifications tab below.