Learn More
Nova Biomedical Corporation Lactate Plus Meter, Rapid Blood Lactate Measurement, Ergonomic Design, 1 meter
Catalog No. NC2543494
Encompass
The Lactate Plus Meter by Nova Biomedical offers swift and precise blood lactate measurements, ideal for athletes and fitness professionals. With a small sample volume of 0.7 µL, it ensures less painful testing without the hassle of calibration codes. This handheld device delivers lab-like accuracy in just 13 seconds, making it a top choice for performance assessment and aerobic conditioning.
Catalog No. NC2543494 Supplier Nova Biomedical Corporation Supplier No. 62624
The Lactate Plus Meter by Nova Biomedical offers swift and precise blood lactate measurements, ideal for athletes and fitness professionals. With a small sample volume of 0.7 µL, it ensures less painful testing without the hassle of calibration codes. This handheld device delivers lab-like accuracy in just 13 seconds, making it a top choice for performance assessment and aerobic conditioning.