Learn More
Waters Corp Needle Kit, 30μL, MP35N, With Guide and Seat
Catalog No. 502391456 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This needle kit provides a replacement needle FTN-30μL MP35N with guide and seat. It is the standard needle assembly used in an Arc Bio Sample Manager-FTN.
  • Standard needle assembly for Arc Bio Sample Manager-FTN
  • 30μL capacity
  • Constructed from MP35N material
  • Includes guide and seat
  • Designed for UPLC system types
  • Annual replacement schedule
Catalog No. 50-239-1456 Supplier Waters Corp Supplier No. 700011760
This needle kit provides a replacement needle FTN-30μL MP35N with guide and seat. It is the standard needle assembly used in an Arc Bio Sample Manager-FTN.
  • Standard needle assembly for Arc Bio Sample Manager-FTN
  • 30μL capacity
  • Constructed from MP35N material
  • Includes guide and seat
  • Designed for UPLC system types
  • Annual replacement schedule