Learn More
Waters Corp Waters O-Ring, Ion Block, FKM, AS016, 10 per Pack
Catalog No. 50819193 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement O-ring, designated AS016, is designed for use in the source assemblies of various systems, including 3100, SQD, SQD2, ACQUITY TQD, Xevo TQD, and Legacy ESI Probes.
  • Made from FKM material
  • Suitable for MS system type
  • Comes in a package of 10
  • Recommended for replacement as required
Catalog No. 50-819-193 Supplier Waters Corp Supplier No. 700000355
This replacement O-ring, designated AS016, is designed for use in the source assemblies of various systems, including 3100, SQD, SQD2, ACQUITY TQD, Xevo TQD, and Legacy ESI Probes.
  • Made from FKM material
  • Suitable for MS system type
  • Comes in a package of 10
  • Recommended for replacement as required