Learn More
Electron Microscopy Sciences Forensic Collection Kit with Plastic Vial
SDP
Supplier:  Electron Microscopy Sciences 7648003
Encompass_Preferred
These kits are suitable for field work and gunshot residue in the lab or out. Manufactured from a special certified aluminum alloy, containing only trace amounts of Barium (~0.00010%), Copper (~0.0139%), Zinc (~0.0094%), Nickel (~0.00309%), and Antimony (~0.00080%). These sample mounts are suitable for typical forensic sample studies. The kit consists of 10 pin mounts (12.74 mm diameter) 10 carbon adhesive tabs, 10 storage vials, and one plastic tweezer.
Catalog No. 50-245-90