Learn More
Chemglass Life Sciences Kit, Clear with Spot 2.0mL 12x32mm 9mm Thread Large Opening Vial and 0.040" PTFE/Red Rubber Septa Lined Blue Closure
SDP
Catalog No. 501532648
Encompass_Preferred
Kit, Clear with Spot 2.0mL 12x32mm 9mm Thread Large Opening Vial and 0.040" PTFE/Red Rubber Septa Lined Blue Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Catalog No. 50-153-2648 Supplier Chemglass Life Sciences Supplier No. CV52511232
Kit, Clear with Spot 2.0mL 12x32mm 9mm Thread Large Opening Vial and 0.040" PTFE/Red Rubber Septa Lined Blue Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.