Learn More
Chemglass Life Sciences Kit, Clear 2.0mL 12x32mm GPI 10-425 Large Opening Screw Thread Vial and 0.065" Red PTFE/Silicone Septa Lined Closure
SDP
Catalog No. 501532645
Encompass_Preferred
Kit, Clear 2.0mL 12x32mm GPI 10-425 Large Opening Screw Thread Vial and 0.065" Red PTFE/Silicone Septa Lined Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Catalog No. 50-153-2645 Supplier Chemglass Life Sciences Supplier No. CV52251232
Kit, Clear 2.0mL 12x32mm GPI 10-425 Large Opening Screw Thread Vial and 0.065" Red PTFE/Silicone Septa Lined Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.