Learn More
Chemglass Life Sciences Kit, Amber 2.0mL 12x32mm GPI 10-425 Large Opening Screw Thread Vial and 0.010" PTFE Septa Lined Closure
SDP
Catalog No. 501532617
Encompass_Preferred
Kit, Amber 2.0mL 12x32mm GPI 10-425 Large Opening Screw Thread Vial and 0.010" PTFE Septa Lined Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Catalog No. 50-153-2617 Supplier Chemglass Life Sciences Supplier No. CV50151232
Kit, Amber 2.0mL 12x32mm GPI 10-425 Large Opening Screw Thread Vial and 0.010" PTFE Septa Lined Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.