Learn More
Chemglass Life Sciences Kit, Clear 4.0mL 15x45mm GPI 13-425 WISP Vial and 0.010" PTFE Septa Lined Closure
SDP
Catalog No. 50231223
Encompass_Preferred
Kit, Clear 4.0mL 15x45mm GPI 13-425 WISP Vial and 0.010" PTFE Septa Lined Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.
Catalog No. 50-231-223 Supplier Chemglass Life Sciences Supplier No. CV50001545
Kit, Clear 4.0mL 15x45mm GPI 13-425 WISP Vial and 0.010" PTFE Septa Lined Closure. These convenience kits come with 100 vials with the closures attached. Packaged in convenient vial trays.