Learn More
Perkin Elmer US LLC PYRIS 1 TGA Hangdown Wire Kit - Qty. 5
Catalog No. 509052222
Encompass_Preferred
Hangdown wires suspend down from the TGA Furnace Balance Mechanism and catch a sample pan's stirrup and lift the pan from the Autosampler and into the furnace. The hangdown wires come in a variety of materials and kit sizes.
Catalog No. 50-905-2222 Supplier Perkin Elmer US LLC Supplier No. N5370487
Add to Cart
Add to Cart
Hangdown wires suspend down from the TGA Furnace Balance Mechanism and catch a sample pan's stirrup and lift the pan from the Autosampler and into the furnace. The hangdown wires come in a variety of materials and kit sizes.