Learn More
Thermo Scientific Kit Rebuild Vlv 6-Port 6K 1Ea
Supplier:  Thermo Scientific 075973
Encompass
valve rebuild kit for 6-port 6-KG capacity valve contains necessary parts & components to refurbish worn or damaged 6-port 6-KG valve restoring proper valve operation & performance kit likely includes valve seats seals springs & other small valve components that commonly require replacement after extended use
Catalog No. 50-134-5981