Learn More
Waters Corp Waters Rebuild Kit, Reference Valve, HPLC, Annual
Catalog No. 50819018 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Rebuild Kit offers a cost-effective solution for maintaining a pump’s reference valve. It includes all necessary parts and instructions for rebuilding the valve, which is recommended for annual replacement or rebuilding. It is suitable for HPLC systems.
  • Provides all necessary parts and instructions to rebuild the reference valve.
  • Contains a diaphragm.
  • Contains an actuator.
  • Includes a valve button.
  • Includes a gasket.
  • Recommended for annual replacement or rebuilding.
Catalog No. 50-819-018 Supplier Waters Corp Supplier No. WAT025746
This Rebuild Kit offers a cost-effective solution for maintaining a pump’s reference valve. It includes all necessary parts and instructions for rebuilding the valve, which is recommended for annual replacement or rebuilding. It is suitable for HPLC systems.
  • Provides all necessary parts and instructions to rebuild the reference valve.
  • Contains a diaphragm.
  • Contains an actuator.
  • Includes a valve button.
  • Includes a gasket.
  • Recommended for annual replacement or rebuilding.