Learn More
Hach Company KIT - Salt Bridge-Hi-Purity
Supplier:  Hach Company 609765009001
Encompass
Field replaceable salt bridge has a ceramic inner junction PVDF outer junction FKM/FPM O-Ring and contains salt solution Only for use with LCP 1-1/2 Inch differential pH sensors
Catalog No. 50-002-07203