Learn More
TA Instruments Kit Sample Pans Quartz Pkg of 3, 180µL Volume, for Discovery SA, Reusable with Proper Handling
Catalog No. 5000580739 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Kit Sample Pans Quartz Pkg Of 3 consists of TGA quartz sample pans with 180 µL volume designed primarily for use with the Discovery SA. Quartz sample pans may be reused for subsequent analyses with proper handling and cleaning.
Catalog No. 50-005-80739 Supplier TA Instruments Supplier No. 957210.902
Waters TA Instruments Kit Sample Pans Quartz Pkg Of 3 consists of TGA quartz sample pans with 180 µL volume designed primarily for use with the Discovery SA. Quartz sample pans may be reused for subsequent analyses with proper handling and cleaning.