Learn More
Med Vet International 48-Pc. Advanced Skin Stapler Wound Closure Surgical Kit
Catalog No. 502829940
Encompass_Preferred
This all-inclusive kits provides the essential first aid for cleaning and flushing a wound, skin prepping, wound closure, as well as post-op supplies for after. If you are looking for an all in one wound closure solution to add to your first aid supplies, this may be the kit for you. Features Saline Wound Flush Sterile Surgical Skin Stapler with Surgical Staple Remover Staple Remover Medical Grade Instrument Sterile Gauze Sponges & Roll Sterile Surgical Drape Bacitracin OintmentContents1 - Sterile Surgical Skin Stapler, 35 Wide Staples1 - Staple Remover4 - 2.5mL Saline Wound Flush Syringe2 - Exam Gloves8 - Sterile 4x4 Gauze, 12 Ply, Sterile 2 Pack1 - Sterile Surgical Drape, 18x26in1 - Adson Tissue Forcep, 4.5in 1x2 Teeth4 - Bacitracin 0.9G Foil Pk4 - Sterile Stretch Gauze Rolls 4in X 2.1 yds4 - 1"x1.5yds Plastic Tape1 - Cohesive Bandage 4in x 5yds8 - 70% Alcohol Prep Pads8 - Povidone Iodine Prep Pad1 - Sterile Abdominal Pad 8 x 7.5in
Catalog No. 50-282-9940 Supplier Med Vet International Supplier No. SHS7
 Restricted Item
Add to Cart
Add to Cart
This all-inclusive kits provides the essential first aid for cleaning and flushing a wound, skin prepping, wound closure, as well as post-op supplies for after. If you are looking for an all in one wound closure solution to add to your first aid supplies, this may be the kit for you. Features Saline Wound Flush Sterile Surgical Skin Stapler with Surgical Staple Remover Staple Remover Medical Grade Instrument Sterile Gauze Sponges & Roll Sterile Surgical Drape Bacitracin OintmentContents1 - Sterile Surgical Skin Stapler, 35 Wide Staples1 - Staple Remover4 - 2.5mL Saline Wound Flush Syringe2 - Exam Gloves8 - Sterile 4x4 Gauze, 12 Ply, Sterile 2 Pack1 - Sterile Surgical Drape, 18x26in1 - Adson Tissue Forcep, 4.5in 1x2 Teeth4 - Bacitracin 0.9G Foil Pk4 - Sterile Stretch Gauze Rolls 4in X 2.1 yds4 - 1"x1.5yds Plastic Tape1 - Cohesive Bandage 4in x 5yds8 - 70% Alcohol Prep Pads8 - Povidone Iodine Prep Pad1 - Sterile Abdominal Pad 8 x 7.5in