Learn More
Beckman Coulter Microfuge Tubes, 1.5mL, Natural, Bag of 1000
Catalog No. NC1487509
Encompass
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop microcentrifuge designed for quick spin-downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 xg. The rotor accommodates 18 x 1.5/2ml microcentrifuge tubes. It has a compact footprint, digital display, 10 acceleration/deceleration rates, and a 30 minute timer with hold function. The Microfuge 18 is designed for routine applications in life science laboratories.
This item is not returnable. View return policy
Catalog No. NC1487509 Supplier Beckman Coulter Supplier No. 208900601
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop microcentrifuge designed for quick spin-downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 xg. The rotor accommodates 18 x 1.5/2ml microcentrifuge tubes. It has a compact footprint, digital display, 10 acceleration/deceleration rates, and a 30 minute timer with hold function. The Microfuge 18 is designed for routine applications in life science laboratories.