Learn More
Molecular Devices Molecular Devices Reporter Assay Kit, Dual Luciferase, 1,000 Reactions
Catalog No. 5000014583
Encompass_Preferred
SpectraMax DuoLuc Reporter Assay Kit enables sensitive, quantitative measurement of firefly and Renilla luciferase activities in mammalian cells using flash-type luminescence. The bulk format supports high-throughput workflows and is optimized for use with microplate readers equipped with injectors. Preconfigured reader protocols simplify data acquisition and normalization to internal controls.
  • Dual luciferase assay for firefly and Renilla measurements.
  • Bulk kit provides reagents for 1,000 reactions.
  • Flash-type luminescence chemistry for high sensitivity.
  • Optimized for microplate readers with injector capability.
  • Enables normalization to an internal control reporter.
  • Includes assay reagents; additional passive lysis buffer available separately.
Catalog No. 50-000-14583 Supplier Molecular Devices Supplier No. R8362
SpectraMax DuoLuc Reporter Assay Kit enables sensitive, quantitative measurement of firefly and Renilla luciferase activities in mammalian cells using flash-type luminescence. The bulk format supports high-throughput workflows and is optimized for use with microplate readers equipped with injectors. Preconfigured reader protocols simplify data acquisition and normalization to internal controls.
  • Dual luciferase assay for firefly and Renilla measurements.
  • Bulk kit provides reagents for 1,000 reactions.
  • Flash-type luminescence chemistry for high sensitivity.
  • Optimized for microplate readers with injector capability.
  • Enables normalization to an internal control reporter.
  • Includes assay reagents; additional passive lysis buffer available separately.