Learn More
Molecular Devices Molecular Devices Deep Well Plate, Polypropylene, 2,000 μl, 96-Well, Square
Catalog No. 5000014341
Encompass_Preferred
Molecular Devices 96-well deep well plate provides high-volume sample storage and processing with a 2,000 μl working volume per well. The polypropylene plate features square wells and elevated well walls for easier sealing and handling, and it conforms to SBS microplate dimensions. Lids and cap mats are available separately.
  • 96-well format
  • Working volume 2,000 μl per well
  • Square well format
  • Material polypropylene
  • Conforms to SBS footprint and dimensions
  • Packed in sleeves of 6, 24 plates per case
Catalog No. 50-000-14341 Supplier Molecular Devices Supplier No. S1205
Molecular Devices 96-well deep well plate provides high-volume sample storage and processing with a 2,000 μl working volume per well. The polypropylene plate features square wells and elevated well walls for easier sealing and handling, and it conforms to SBS microplate dimensions. Lids and cap mats are available separately.
  • 96-well format
  • Working volume 2,000 μl per well
  • Square well format
  • Material polypropylene
  • Conforms to SBS footprint and dimensions
  • Packed in sleeves of 6, 24 plates per case