Learn More
Thermo Scientific KIT STNDRD AUTSMPLRVIAL 100/PK
Catalog No. 50465242 Shop All Thermo Scientific Products
Encompass
automsampler vial kits. 1.8 ML SCREW CaP CLEaR - 12MM X 32MM
Catalog No. 50-465-242 Supplier Thermo Scientific Supplier No. A4954010
automsampler vial kits. 1.8 ML SCREW CaP CLEaR - 12MM X 32MM