Learn More
Med Vet International 13-pc. Emergency Wound Care Suture Kit - Minimalist Edition
Catalog No. 502522174
Encompass_Preferred
This 13 pc minimalist suture kit will provide you with the basics for suturing. It provides all necessary tools for suturing, with no extra bulk. Features Sterile in-date sutures of the most common types Medical grade quality instruments Content (1) 4-0 Sterile Suture with Needle (1) #3 Scalpel Handle, Fits Blades #10-#15 (2) #10 Sterile Scalpel Blades (2) #11 Sterile Scalpel Blades (1) 5in Needle Holder or larger (size varies) (1) 5in Curved Hemostat or larger (size varies) (1) 4.5in OR Scissors (2) Alcohol Prep Pads, 70% Isopropyl Antiseptic (2) Povidone Iodine Prep Pads Antiseptic Carrying Case All instruments are surgical stainless steel. Sutures are in-date, sterile. When it counts, this lightweight and compact suture kit is easily deployable to treat most wounds.
Catalog No. 50-252-2174 Supplier Med Vet International Supplier No. SK1
This 13 pc minimalist suture kit will provide you with the basics for suturing. It provides all necessary tools for suturing, with no extra bulk. Features Sterile in-date sutures of the most common types Medical grade quality instruments Content (1) 4-0 Sterile Suture with Needle (1) #3 Scalpel Handle, Fits Blades #10-#15 (2) #10 Sterile Scalpel Blades (2) #11 Sterile Scalpel Blades (1) 5in Needle Holder or larger (size varies) (1) 5in Curved Hemostat or larger (size varies) (1) 4.5in OR Scissors (2) Alcohol Prep Pads, 70% Isopropyl Antiseptic (2) Povidone Iodine Prep Pads Antiseptic Carrying Case All instruments are surgical stainless steel. Sutures are in-date, sterile. When it counts, this lightweight and compact suture kit is easily deployable to treat most wounds.