Learn More
Azenta US Inc KIT TAPE 5 SPOOL
Catalog No. NC9035571
Encompass
  • Product type: Removal tape
  • Compatibility: Azenta Automated Plate Seal Remover (XPeel)
  • Kit quantity: 5 rolls
  • Removals per roll: Up to 400 seals
  • Seal compatibility: Heat and pressure applied seals
  • Microplate compatibility: Full-skirted PCR plates, low-base microplates, deep-well plates (up to 2 ml)
  • Benefit: Reduces cross-contamination
  • Benefit: Preserves sample integrity
  • Benefit: Eliminates manual de-sealing
  • Integration: Manual or automated systems with external robotics
Catalog No. NC9035571 Supplier Azenta US Inc Supplier No. XTAPE_2000
  • Product type: Removal tape
  • Compatibility: Azenta Automated Plate Seal Remover (XPeel)
  • Kit quantity: 5 rolls
  • Removals per roll: Up to 400 seals
  • Seal compatibility: Heat and pressure applied seals
  • Microplate compatibility: Full-skirted PCR plates, low-base microplates, deep-well plates (up to 2 ml)
  • Benefit: Reduces cross-contamination
  • Benefit: Preserves sample integrity
  • Benefit: Eliminates manual de-sealing
  • Integration: Manual or automated systems with external robotics