Learn More
Seracare Life Sciences Inc TMB 2-Component Microwell Peroxidase Substrate Kit 600mL
Catalog No. 5067421
Encompass_Preferred
Most sensitive chromogenic substrate for peroxidase detection, more sensitive than ABTS and OPD. It develops a deep blue color in the presence of peroxidase-labeled conjugates measurable at 650 nm. Turns yellow when stopped by acidification and read at 450 nm. Recommended for ELISA. TMB can also be used in blotting applications with the purchase of the TMB Membrane Enhancer (50-77-01). 2-component format of 3, 3', 5, 5' - Tetramethylbenzidine, simply mix and use. Has longer shelf life than 1-component TMB substrates.
Item requires ice pack and is temperature sensitive.
This item is not returnable. View return policy
Catalog No. 50-674-21 Supplier Seracare Life Sciences Inc Supplier No. 51200047
Most sensitive chromogenic substrate for peroxidase detection, more sensitive than ABTS and OPD. It develops a deep blue color in the presence of peroxidase-labeled conjugates measurable at 650 nm. Turns yellow when stopped by acidification and read at 450 nm. Recommended for ELISA. TMB can also be used in blotting applications with the purchase of the TMB Membrane Enhancer (50-77-01). 2-component format of 3, 3', 5, 5' - Tetramethylbenzidine, simply mix and use. Has longer shelf life than 1-component TMB substrates.
Item requires ice pack and is temperature sensitive.