Learn More
Kuhner Shaker Inc KIT TO CONVERT STANDARD WATER
Catalog No. 502570320
Encompass
Kit to convert standard water bath for use with Kuhner's CleanShield Technology. Compatible with Kuhner Shaker's ISF1-Z Series Shaker Incubators with Humidity Control Capability only
Catalog No. 50-257-0320 Supplier Kuhner Shaker Inc Supplier No. NSMZ856187
Kit to convert standard water bath for use with Kuhner's CleanShield Technology. Compatible with Kuhner Shaker's ISF1-Z Series Shaker Incubators with Humidity Control Capability only