Learn More
TA Instruments Kit Tzero Calibration Sapphire, Two Sapphire Discs, Tzero Calibration of DSC Cell, Accurate Temperature Measurements, Reliable DSC Performance
Catalog No. 5000580900 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Kit Tzero Calibration Sapphire contains two sapphire discs used for Tzero calibration of the DSC cell, ensuring accurate temperature measurements and reliable DSC performance.
Catalog No. 50-005-80900 Supplier TA Instruments Supplier No. 970345.901
Waters TA Instruments Kit Tzero Calibration Sapphire contains two sapphire discs used for Tzero calibration of the DSC cell, ensuring accurate temperature measurements and reliable DSC performance.