Learn More
Waters Corp Union Kit, Differential Diameter, 4 mm to 6 mm
Catalog No. 50792397 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This union kit is designed for connecting tubing with differential diameters. It facilitates connections between 4 mm and 6 mm tubing, ensuring secure and reliable seals in various applications.
  • Designed for differential diameter connections
  • Connects 4 mm and 6 mm tubing
  • Ensures secure and reliable seals
Catalog No. 50-792-397 Supplier Waters Corp Supplier No. 700000465
This union kit is designed for connecting tubing with differential diameters. It facilitates connections between 4 mm and 6 mm tubing, ensuring secure and reliable seals in various applications.
  • Designed for differential diameter connections
  • Connects 4 mm and 6 mm tubing
  • Ensures secure and reliable seals