Learn More
Hach Company Kit-Union Mounting-316SS
Supplier:  Hach Company 6131400
Encompass
316 stainless steel union mount for Hach pHD sensor Includes standard 1 5in tee special union pipe with adapter sealing hub lock ring in CPVC and FKM/FPM O-ring
Catalog No. 50-002-07228