Learn More
Waters Corp Kit, Upgrade, ACQUITY UPLC Local Console Controller
Catalog No. 50791693 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC Local Console Controller Upgrade Kit includes the Y-cable and an external Power Supply for controlling an existing ACQUITY Local Console Controller via a non-powered ethernet port. The ACQUITY Local Console Controller has now been discontinued.
Catalog No. 50-791-693 Supplier Waters Corp Supplier No. 205000590
The ACQUITY UPLC Local Console Controller Upgrade Kit includes the Y-cable and an external Power Supply for controlling an existing ACQUITY Local Console Controller via a non-powered ethernet port. The ACQUITY Local Console Controller has now been discontinued.