Learn More
Chemglass Life Sciences Vacuum Adapter Kit, Supplied with (1) Polypropylene Box and (1) each of the following: CG-1049-C-02, CG-1049-C-04, CG-1049-C-06, CG-1049-C-50, CG-1401-11
SDP
Catalog No. 50872882
Encompass_Preferred
Vacuum Adapter Kit, Supplied with (1) Polypropylene Box and (1) each of the following: CG1049C02, CG1049C04, CG1049C06, CG1049C-50, CG140111. Adapters are used for filtration directly into vials. There are three top configurations, with the bottom of all adapters having a 20-400 GPI thread for connection to vials using a CG-1318 Threaded Connector. Design incorporates a drip tube sealed within thread to prevent breakage & are supplied with a 2-1/2¿ PTFE delivery stem. Hose connection is 10mm OD at the largest serration. ADAPTERS ARE SUPPLIED WITH PTFE DELIVERY STEM.¿ Adapter with tooled flare top is used for reduced pressure filtration with plain stem Buchner funnels. Flare accepts a #2 Pluro Stopper. ¿ Adapter with outer luer joint provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes or syringes.¿ Adapter with 20-400 GPI thread allows for transfer from vial to vial.
Catalog No. 50-872-882 Supplier Chemglass Life Sciences Supplier No. CG1049C10
Vacuum Adapter Kit, Supplied with (1) Polypropylene Box and (1) each of the following: CG1049C02, CG1049C04, CG1049C06, CG1049C-50, CG140111. Adapters are used for filtration directly into vials. There are three top configurations, with the bottom of all adapters having a 20-400 GPI thread for connection to vials using a CG-1318 Threaded Connector. Design incorporates a drip tube sealed within thread to prevent breakage & are supplied with a 2-1/2¿ PTFE delivery stem. Hose connection is 10mm OD at the largest serration. ADAPTERS ARE SUPPLIED WITH PTFE DELIVERY STEM.¿ Adapter with tooled flare top is used for reduced pressure filtration with plain stem Buchner funnels. Flare accepts a #2 Pluro Stopper. ¿ Adapter with outer luer joint provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes or syringes.¿ Adapter with 20-400 GPI thread allows for transfer from vial to vial.