Learn More
Chemglass Life Sciences Kit, Vial, Autosampler HPLC Vial, 0.5mL, Plastic, PP, Limited Volume, Large Opening, 9mm Thread, 12 x 32mm, with 0.30mL Fused Insert, Blue Bonded Closure, Open Hole, .040" PTFE/Silicone Septa with Slit
SDP
Catalog No. 501532640
Encompass_Preferred
Kit, Preassembled Vial, Autosampler HPLC Vial, 0.5mL, RAM, Plastic, PP, Limited Volume, Large Opening, 9mm Thread, 12 x 32mm, with 0.30mL Fused Insert, Blue Bonded Closure, Open Hole, .040" PTFE/Silicone Septa with Slit. These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.
Catalog No. 50-153-2640 Supplier Chemglass Life Sciences Supplier No. CV51861232
Kit, Preassembled Vial, Autosampler HPLC Vial, 0.5mL, RAM, Plastic, PP, Limited Volume, Large Opening, 9mm Thread, 12 x 32mm, with 0.30mL Fused Insert, Blue Bonded Closure, Open Hole, .040" PTFE/Silicone Septa with Slit. These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.