Learn More
Sigma Aldrich Fine Chemicals Biosciences KitAlysis High-Throughput 24 Pd Precatalyst Reaction Screening Kit-Pack of 2
Catalog No. NC1585498
Encompass
Screening kit for cross-coupling reactions. Each kit contains 2 customizable screening sets that include the following components: 24 pre-weighed catalysts in glass micro vials loaded with stir bars and topped with cap mat. 2 KitAlysis 24-Well reaction block replacement films. 4 empty substrate stock solution reaction vials with stir bars. 3 stir bars for stock solutions. 1 internal standard.
Catalog No. NC1585498 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. KITALYSIS24PD2PK
Add to Cart
Add to Cart
Screening kit for cross-coupling reactions. Each kit contains 2 customizable screening sets that include the following components: 24 pre-weighed catalysts in glass micro vials loaded with stir bars and topped with cap mat. 2 KitAlysis 24-Well reaction block replacement films. 4 empty substrate stock solution reaction vials with stir bars. 3 stir bars for stock solutions. 1 internal standard.