Learn More
Sigma Aldrich Fine Chemicals Biosciences KitAlysisTM Benchtop Inert1EA
Catalog No. 502909001
Encompass
Provides an inert environment to run oxygen sensitive cross-coupling reactions in a laboratory fume hood.Designed to be used with KitAlysis High-Throughput Screening Kits and KitAlysis 24-Well Reaction Block.
Catalog No. 50-290-9001 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7420641EA
Provides an inert environment to run oxygen sensitive cross-coupling reactions in a laboratory fume hood.Designed to be used with KitAlysis High-Throughput Screening Kits and KitAlysis 24-Well Reaction Block.