Learn More
World Precision Instrument Kite Manual Micromanipulator-KITE-R
SDP
Catalog No. NC9661462
Encompass
The KITE is an economical manual micromanipulator that is perfect for student use. It is available in right or left hand versions and is also sold bundled with an optional M3 Tilting Base. Our 5 lb. weight (WPI #5464) is frequently sold with this unit to provide stability.
Catalog No. NC9661462 Supplier World Precision Instrument Supplier No. KITER
The KITE is an economical manual micromanipulator that is perfect for student use. It is available in right or left hand versions and is also sold bundled with an optional M3 Tilting Base. Our 5 lb. weight (WPI #5464) is frequently sold with this unit to provide stability.