Learn More
Med Vet International EasyTouch HealthPro Glucose Meter Monitoring System Kit
Supplier:  Med Vet International 809001
Encompass_Preferred
The EasyTouch HealthPro™ Glucose Monitoring System gives you the ability to obtain a blood sample for testing your sugar from different areas of your body. You may test your blood sugar from your forearm, upper arm, palm (ventral or dorsal hand), thigh, calf or fingertips. Glucose Meter Contents EasyTouch HealthPro™ Meter One Lancing Device Ten Twist Lancets One Deluxe Carrying Case User Manual Log Book Quick Reference Guide Features & Benefits 100% Code Free Test Strip Ejector Backlit Screen Tiny Blood Sample Required Only 0.5 L 5 Second Rapid Test Results Alternate Site Testing Status of User Condition (Activity, Medication & Meal Recognition) 365 Result History Memory True Lifetime Warranty CLIA Certified 7,14, & 21 Day Averaging
Catalog No. 50-255-9667