Learn More
Med Vet International EasyTouch Glucose Monitoring System Kit, Each
Catalog No. 502559672
Encompass_Preferred
The EasyTouch Glucose Monitoring System gives you the ability to obtain a blood sample for testing your sugar from different areas of your body. You may test your blood sugar from your forearm, palm (ventral or dorsal hand), or fingertips. Glucose Meter Contents EasyTouch Meter One Lancing Device Ten Twist Lancets One Deluxe Carrying Case User Manual Log Book Quick Reference Guide Features & Benefits 100% Code Free Tiny Blood Sample Required Only 0.5 L 5 Second Rapid Test Results Alternate Site Testing 300 Result History Memory 7,14, & 30 Day Averaging 4 Alarm Settings Auto On/Off True Lifetime Warranty CLIA Certified 7,14, & 30 Day Averaging 4 Alarm Settings Auto On/Off True Lifetime Warranty CLIA Certified
Catalog No. 50-255-9672 Supplier Med Vet International Supplier No. 807001
The EasyTouch Glucose Monitoring System gives you the ability to obtain a blood sample for testing your sugar from different areas of your body. You may test your blood sugar from your forearm, palm (ventral or dorsal hand), or fingertips. Glucose Meter Contents EasyTouch Meter One Lancing Device Ten Twist Lancets One Deluxe Carrying Case User Manual Log Book Quick Reference Guide Features & Benefits 100% Code Free Tiny Blood Sample Required Only 0.5 L 5 Second Rapid Test Results Alternate Site Testing 300 Result History Memory 7,14, & 30 Day Averaging 4 Alarm Settings Auto On/Off True Lifetime Warranty CLIA Certified 7,14, & 30 Day Averaging 4 Alarm Settings Auto On/Off True Lifetime Warranty CLIA Certified