Learn More
ACROBiosystems ELISA Buffer Set-96T
Catalog No. 502019627
Encompass_Preferred
Kits;96TESTS;This ELISA Buffer Set-96T contains a High-bind 96well plate, coating buffer, washing buffer, blocking buffer, substrate solution and stop solution. The ELISA Buffer Set-96T can be quickly applied to the ELISA experiment and save your time to prepare buffers. This ELISA Buffer Set-96T is sterile-filtered.
Catalog No. 50-201-9627 Supplier ACROBiosystems Supplier No. EBS00196TESTS
Kits;96TESTS;This ELISA Buffer Set-96T contains a High-bind 96well plate, coating buffer, washing buffer, blocking buffer, substrate solution and stop solution. The ELISA Buffer Set-96T can be quickly applied to the ELISA experiment and save your time to prepare buffers. This ELISA Buffer Set-96T is sterile-filtered.