Learn More
Thermo Scientific Thermostat Kit 2pk
Catalog No. 501347653 Shop All Thermo Scientific Products
Encompass
Kit contains support ring and piston seal for repairing and maintaining piston seal assembly in pneumatic, hydraulic and reciprocating pump applications; provides tight dynamic seal between piston and cylinder bore allowing smooth linear movement.
Catalog No. 50-134-7653 Supplier Thermo Scientific Supplier No. 6025.2011A
Kit contains support ring and piston seal for repairing and maintaining piston seal assembly in pneumatic, hydraulic and reciprocating pump applications; provides tight dynamic seal between piston and cylinder bore allowing smooth linear movement.