Learn More
Medchemexpress LLC HY-19735 10mg , KJ Pyr 9 CAS:581073-80-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1973510MG
Encompass_Preferred
HY-19735 10mg KJ Pyr 9 CAS: 581073-80-5 KJ Pyr 9 is an inhibitor of MYC with a Kd of 6.5 nM in in vitro assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9561