Learn More
TARGETMOL CHEMICALS INC KJ PYR 9 25MG
SDP
Catalog No. 502581211
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KJ Pyr 9 is an MYC inhibitor (Kd 6.5 nM in vitro assay). purity: 98%
Catalog No. 50-258-1211 Supplier TARGETMOL CHEMICALS INC Supplier No. T1566525MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KJ Pyr 9 is an MYC inhibitor (Kd 6.5 nM in vitro assay). purity: 98%