Learn More
Medchemexpress LLC KJ Pyr 9 | 581073-80-5 | 99.6% | 385.37 | 5 MG
SDP
Catalog No. 5000393396
Encompass_Preferred
KJ Pyr 9 is an inhibitor of MYC, interfering with MYC-MAX complex formation and inhibiting oncogenic transformation. It reduces the MYC-driven transcriptional signature and inhibits the proliferation of various cell lines, with higher sensitivity for Burkitt lymphoma cell lines. In vivo, it demonstrates the ability to halt tumor growth.
  • Interferes with MYC-MAX complex formation
  • Inhibits MYC-induced oncogenic transformation in cell culture
  • Reduces the MYC-driven transcriptional signature
  • Inhibits proliferation of NCI-H460, MDA-MB-231, and SUM-159PT cell lines
  • More sensitive for Burkitt lymphoma cell lines with high c-MYC expression
  • Halts tumor growth in vivo at 10 mg/kg
Catalog No. 50-003-93396 Supplier Medchemexpress LLC Supplier No. 5MGHY19735
Add to Cart
Add to Cart
KJ Pyr 9 is an inhibitor of MYC, interfering with MYC-MAX complex formation and inhibiting oncogenic transformation. It reduces the MYC-driven transcriptional signature and inhibits the proliferation of various cell lines, with higher sensitivity for Burkitt lymphoma cell lines. In vivo, it demonstrates the ability to halt tumor growth.
  • Interferes with MYC-MAX complex formation
  • Inhibits MYC-induced oncogenic transformation in cell culture
  • Reduces the MYC-driven transcriptional signature
  • Inhibits proliferation of NCI-H460, MDA-MB-231, and SUM-159PT cell lines
  • More sensitive for Burkitt lymphoma cell lines with high c-MYC expression
  • Halts tumor growth in vivo at 10 mg/kg