Learn More
Medchemexpress LLC KJ Pyr 9 | 581073-80-5 | 99.6% | 385.37 | 50 MG
SDP
Catalog No. 5000364489
Encompass_Preferred
KJ Pyr 9 is an inhibitor of MYC with a Kd of 6.5 nM in in vitro assay. It interferes with MYC-MAX complex formation in cells and specifically inhibits MYC-induced oncogenic transformation in cell culture. It also preferentially interferes with the proliferation of MYC-overexpressing human and avian cells.
  • Inhibits MYC-MAX complex formation
  • Inhibits MYC-induced oncogenic transformation
  • Reduces MYC-driven transcriptional signature
  • Inhibits proliferation of NCI-H460, MDA-MB-231, and SUM-159PT cell lines
  • Halts tumor growth in vivo
Catalog No. 50-003-64489 Supplier Medchemexpress LLC Supplier No. 50MGHY19735
Add to Cart
Add to Cart
KJ Pyr 9 is an inhibitor of MYC with a Kd of 6.5 nM in in vitro assay. It interferes with MYC-MAX complex formation in cells and specifically inhibits MYC-induced oncogenic transformation in cell culture. It also preferentially interferes with the proliferation of MYC-overexpressing human and avian cells.
  • Inhibits MYC-MAX complex formation
  • Inhibits MYC-induced oncogenic transformation
  • Reduces MYC-driven transcriptional signature
  • Inhibits proliferation of NCI-H460, MDA-MB-231, and SUM-159PT cell lines
  • Halts tumor growth in vivo