Learn More
Selleck Chemical LLC KJ Pyr 9-E1756-5MG
SDP
Supplier:  Selleck Chemical LLC E17565MG
Encompass_Preferred
KJ Pyr 9 is an inhibitor of MYC with a Kd of 6 5 nM in in vitro assay
Catalog No. 50-313-8939