Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kjeldahl Digestion Solution, Reagents

Catalog No. 01334858
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-858 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-858 Supplier Reagents Holdings Llc Supplier No. CK2001001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kjeldahl Digestion Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Kjeldahl Digestion Solution
Boiling Point 102°C
Color Colorless
CAS 7778-80-5, 7664-93-9, 7732-18-5
Physical Form Liquid
Quantity 1 L
Packaging Poly Bottle
Synonym Nitrogen
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.